missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
FSBA Monoclonal antibody specifically detects FSBA in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Spezifikation
Spezifikation
| Antigen | FSBA |
| Anwendungen | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Klassifikation | Monoclonal |
| Klon | FSBA 2F5/5 |
| Konjugat | Janelia Fluor 549 |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | 5'-(4-(fluorosulfonyl)benzoyl)adenosine, 5'-(4-fluorosulfonylbenzoyl)adenosine, 5'-FSBA, p-fluorosulfonylbenzoyl-5'-adenosine |
| Wirtsspezies | Mouse |
| Immunogen | Fluorylsulponylbenzoyladenosine (FSBA) |
| Reinigungsverfahren | Protein A purified |
| Mehr anzeigen |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?