missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Furin Monoclonal Antibody (222712)
Mouse Monoclonal Antibody
Marke: Invitrogen™ MA560993
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.5 mg/mL in sterile PBS.
Furin is a membrane-associated, calcium-dependent, serine protease that belongs to the subtilisin-like prohormone convertase (PC) family. Members of this family of cellular enzymes cleave most prohormones and neuropeptide precursors. Numerous other cellular proteins, some viral proteins, and bacterial toxins that are transported by the constitutive secretory pathway are also targeted for maturation by PCs. Furin and other PC family members share structural similarities which include a heterogeneous ~10 kDa amino-terminal proregion, a highly conserved ~55 kDa subtilisin-like catalytic domain, and carboxyl-terminal domain that is heterogeneous in length and sequence. These enzymes become catalytically active following proregion cleavage within the appropriate cellular compartment. Furin is the only known PC to possess a transmembrane domain. Cleavage of target proteins occurs at the carboxyl-terminus of the furin consensus sequence, RX(K/R)R. It has been shown that the acidic peptide sequence, C771PSDSEEDEG780, localizes furin to the trans-Golgi-network. Phosphorylation of serine residues within this region modulates intracellular routing of furin protein. An additional signaling domain includes the tetrapeptide sequence, Y759KGL762, which directs internalization from the cell surface.
Spezifikation
| Furin | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P09958 | |
| FURIN | |
| Mouse myeloma cell line NS0-derived recombinant human Furin, Asp108-Glu715. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 222712 | |
| Unconjugated | |
| FURIN | |
| 9130404I01Rik; dibasic processing enzyme; dibasic-processing enzyme; FES upstream region; FUR; Furin; furin (paired basic amino acid cleaving enzyme); furin, membrane associated receptor protein; furin, paired basic amino acid cleaving enzyme; furin; LOW QUALITY PROTEIN: furin; Pace; Paired basic amino acid cleaving enzyme (furin); paired basic amino acid residue cleaving enzyme; paired basic amino acid residue-cleaving enzyme; Pcsk3; prohormone convertase 3; proprotein convertase subtilisin/kexin type 3; Proprotein convertase subtilisin/kexin type 3 (paired basic amino acid cleaving enzyme, furin, membrane associated receptor protein); proprotein convertase subtilisin/kexin type3; SPC1; subtilisin pro-protein processing enzyme | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 5045 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur