missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FZD7 Monoclonal Antibody (151107)

Rat Monoclonal Antibody
Marke: Invitrogen™ MA561077
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD7 protein contains an N-terminal signal sequence, 10 cysteine residues typical of the cysteine-rich extracellular domain of Fz family members, 7 putative transmembrane domains, and an intracellular C-terminal tail with a PDZ domain-binding motif. FZD7 gene expression may downregulate APC function and enhance beta-catenin-mediated signals in poorly differentiated human esophageal carcinomas.
Spécification
| FZD7 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q61090 | |
| FZD7 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-7, Gln33-Leu185 (predicted). | |
| 50 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 151107 | |
| Unconjugated | |
| FZD7 | |
| frizzled 7, seven transmembrane spanning receptor; frizzled class receptor 7; frizzled family receptor 7; frizzled homolog 7; frizzled homolog 7 (Drosophila); Frizzled, drosophila, homolog of, 7; frizzled-7; Fz7; Fz-7; FZD7; FzE3; hFz7; LOC100360552; mFz7; rCG22430-like; rizzled 7 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 14369 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu