Learn More
Gadoxetate Disodium, MedChemExpress
Artikelnummer. 16467008
Encompass_Preferred
Change view
Click to view available options
Menge:
5 mg
10 mg
50 mg
Packungsgröße:
10 Milligramm
5 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16467008 10 mg 10 Milligramm
16463057 50 mg -
16419557 5 mg 5 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16467008 Lieferant Medchem Express Lieferanten-Nr. HY16219
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
MedChemExpress Gadoxetate Disodium (Gd-EOB-DTPA Disodium) is a contrast agent in magnetic resonance imaging (MRI) of the hepatobiliary system, which accumulates in normal, functioning hepatocytes. Gadoxetate Disodium (Gd-EOB-DTPA Disodium) is used to evaluate focal liver lesions, such as hepatocellular carcinoma or liver metastasis on T1-weighted imaging.
Gadoxetate Disodium (Gd-EOB-DTPA Disodium) is a contrast agent in magnetic resonance imaging (MRI) of the hepatobiliary system, which accumulates in normal, functioning hepatocytes. Gadoxetate Disodium (Gd-EOB-DTPA Disodium) is used to evaluate focal liver lesions, such as hepatocellular carcinoma or liver metastasis on T1-weighted imaging.
Spezifikation
Chemischer Name oder Material Gadoxetate Disodium
Physikalische Form Solid
CAS 135326-22-6
Farbe White
Menge 10 mg
Hinweise zur Reinheitsqualität Research
Summenformel C23H28GdN3Na2O11
Zur Verwendung mit (Anwendung) Neuroscience-Neuromodulation
Synonym Gd-EOB-DTPA Disodium ZK 139834
Löslichkeitsinformationen DMSO : 24 mg/mL (33.07 mM; Need ultrasonic)
SMILES O=C1[O-][Gd+3]([N]2(C1)CC3=O)([O-]3)([O-]C4=O)([O-]C5=O)([N]6(C5)C7)([N](C4)(CC2)CC6CC8=CC=C(OCC)C=C8)[O-]C7=O.[Na+].[Na+]
Molekulargewicht (g/mol) 725.71
Formelmasse 725.71
Reinheit (%) 99.0%
Güte Research