Learn More
Invitrogen™ gamma Tubulin Monoclonal Antibody (GT4511)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA531482
Beschreibung
Keep as concentrated solution. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (98%), Xenopus laevis (99%), Dog (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Positive Control: Jurkat, Raji, NCI-H929, Neuro2A, C8D30, NIH-3T3, BCL-1, C2C12, PC-12, Rat-2, mouse testis. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.
Spezifikation
| gamma Tubulin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P23258, P83887, P83888, Q8VCK3, Q9NRH3 | |
| TUBG1, Tubg2 | |
| Recombinant protein encompassing a sequence within the center region of human gamma Tubulin. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Drosophila | |
| Antibody | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| GT4511 | |
| Unconjugated | |
| TUBG1 | |
| 1500010O08Rik; AI451582; AI503389; AI504772; CDCBM4; centrosome marker; gamma tubulin; gamma-1-tubulin; Gamma-2-tubulin; gamma-tubulin; gamma-tubulin complex component 1; GCP-1; similar to tubulin, gamma 1; Tubg; TUBG1; tubg1.L; TUBG2; TUBGCP1; Tubgl; tubulin gamma 1; tubulin gamma 1 L homeolog; tubulin gamma 2; Tubulin gamma-1 chain; Tubulin gamma-2 chain; tubulin, gamma 1; tubulin, gamma 2; tubulin, gamma polypeptide; Unknown (protein for MGC:133882); XELAEV_18002398mg; xgam; zgc:55894 | |
| Mouse | |
| Protein G | |
| RUO | |
| 103733, 103768, 252921, 27175, 680991, 7283 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.