missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAPDH Antibody (CL3265) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-59025UV
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
GAPDH Monoclonal antibody specifically detects GAPDH in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
Spezifikation
| GAPDH | |
| Monoclonal | |
| DyLight 350 | |
| aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH | |
| This antibody was developed using a recombinant protein derived from P04406, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Autophagy, Cancer, Chromatin Research, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control | |
| 2597 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown | |
| CL3265 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur