missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GATA1 Monoclonal Antibody (234737)

Rat Monoclonal Antibody
Marke: Invitrogen™ MA561043
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.5 mg/mL in sterile PBS.
Transcriptional activator or repressor which probably serves as a general switch factor for erythroid development. It binds to DNA sites with the consensus sequence 5'-[AT]GATA[AG]-3' within regulatory regions of globin genes and of other genes expressed in erythroid cells.
Spezifikation
| GATA1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P15976, P17679 | |
| Gata1 | |
| E. coli-derived recombinant human GATA-1, Met1-Ser413. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 234737 | |
| Unconjugated | |
| Gata1 | |
| eryf1; Erythroid transcription factor; erythroid transcription factor 1; GATA binding protein 1; GATA binding protein 1 (globin transcription factor 1); GATA1; GATA-1; GATA-binding factor 1; GATA-binding protein 1; GATA-binding protein 1 (globin transcription factor 1); GF1; Gf-1; globin transcription factor 1; HGNC:4170; NFE1; NF-E1; NF-E1 DNA-binding protein; nuclear factor, erythroid 1; transcription factor GATA1; XLANP; XLTDA; XLTT | |
| Rat | |
| Protein A/G | |
| RUO | |
| 14460, 2623 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur