missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GCLC Recombinant Rabbit Monoclonal Antibody (24GB4860)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557409
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
GCLC Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Glutamate-cysteine ligase, also known as gamma-glutamylcysteine synthetase is the first rate-limiting enzyme of glutathione synthesis. The enzyme consists of two subunits, a heavy catalytic subunit and a light regulatory subunit. This locus encodes the catalytic subunit, while the regulatory subunit is derived from a different gene located on chromosome 1p22-p21. Mutations at this locus have been associated with hemolytic anemia due to deficiency of gamma-glutamylcysteine synthetase and susceptibility to myocardial infarction.
Spezifikation
| GCLC | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Gclc | |
| cb1049; D9Wsu168e; EGK_15023; fe36e11; gamma GCS-HS; gamma-ECS; Gamma-glutamylcysteine synthetase; gamma-glutamylcysteine synthetase heavy subunit; GCL; Gclc; GCS; GCS heavy chain; Ggcs-hs; GLCL; GLCLC; GLCL-H; glutamate-cysteine ligase catalytic subunit; Glutamate--cysteine ligase catalytic subunit; glutamate-cysteine ligase, catalytic subunit; Glutamylcysteine gamma synthetase light chain; wu:fe36e11 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14629, 25283, 2729 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4860 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P19468, P48506, P97494 | |
| Gclc | |
| Recombinant protein of human GCLC (aa 1-300). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur