Learn More
Invitrogen™ GDNF Polyclonal Antibody
GREENER_CHOICE
Goat Polyclonal Antibody
Marke:  Invitrogen™ PA5148442
Artikelnummer. 30535391
  • 560.00 € / 1 Milligramm
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Reconstitute at 1 mg/mL in sterile PBS.
GDNF is a glycosylated, disulfide-bonded homodimer molecule. It was first discovered as a potent survival factor for midbrain dopaminergic neurons and was then shown to rescue these neurons in animal models of Parkinson's disease. GDNF is about 100 times more efficient survival factor for spinal motor neurons than the neurotrophins. GDNF is a neurotrophic factor that enhances survival and morphological differentiation of dopaminergic neurons and increases their high-affinity dopamine uptake.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
GDNF
Polyclonal
Unconjugated
GDNF
AI385739; Astrocyte-derived trophic factor; ATF; ATF1; ATF2; Gdnf; glial cell derived neurotrophic factor; glial cell line derived neurotrophic factor; Glial cell line derived neutrophic factor; glial cell line-derived neurotrophic factor; gndf; HFB1-GDNF; hGDNF; H-GDNF; HSCR3; mGDNF; neurotrophic factor
Goat
Protein A/G
RUO
25453, 2668
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Neutralization, Western Blot
1 mg/mL
PBS with trehalose and no preservative
P39905, Q07731
GDNF
E. coli-derived recombinant human GDNF, Arg109-Ile211.
1 mg
Primary
Human, Rat
Antibody
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen