Learn More
Applied Biosystems™ GeneChip™ Mouse Genome 430 2.0 Array
The GeneChip™ Mouse Genome 430 2.0 Array is the first and most comprehensive whole mouse genome expression array
Spezifikation
| Array | Transkriptom-Profiling |
|---|---|
| Format | Array-Kassette |
| Spezies | Maus |
| Produktlinie | GeneChip™ |
Beschreibung
The GeneChip™ Mouse Genome 430 2.0 Array is the first and most comprehensive whole mouse genome expression array:
• Complete coverage of the Mouse Expression Set 430 for analysis of over 39,000 transcripts on a single array
• The Power of the Probe Set - offering multiple independent measurements for each transcript delivering the greatest accuracy and reproducibility of any microarray platform
• More information from every experiment for more complete and efficient analysis
Array Profile
All probe sets represented on the GeneChip™ Mouse Expression Set 430 are included on the GeneChip Mouse Genome 430 2.0 Array. The sequences from which these probe sets were derived were selected from GenBank™, dbEST, and RefSeq. The sequence clusters were created from the UniGene database (Build 107, June 2002) and then refined by analysis and comparison with the publicly available draft assembly of the mouse genome from the Whitehead Institute for Genome Research (MGSC, April 2002).
Instrument and Software Requirements
• GeneChip Scanner 3000 7G, enabled for High-Resolution Scanning*
• GeneChip Operating Software (GCOS) v1.1.1 (includes the GeneChip Scanner 3000 High-Resolution Scanning Patch)
*GeneChip Scanner 3000 High-Resolution Update is standard on all instruments shipped starting in September 2003 with serial number series 502. Previous versions, such as serial number series 501, will require the GeneChip Scanner 3000 High-Resolution Update, Catalog Number 00-0110, to be installed.
Spezifikation
| Transkriptom-Profiling | |
| Array-Kassette | |
| Maus | |
| GeneChip™ |