missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ GET4 Overexpression Lysate

Artikelnummer. 18222002 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Menge:
0,1 mg
Packungsgröße:
0.10 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
18222002 0,1 mg 0.10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18222002 Lieferant R&D Systems™ Lieferanten-Nr. NBL108544
 Eingeschränkter Artikel
Verfügbar ab: 15-12-2026
Zum Warenkorb hinzufügen
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems.

The GET4 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Zelllinie The lysate was created in HEK293T cells, using plasmid ID RC222 and based on accession number NM_15949. The protein contains a C-terminal DDK tag.
Menge 0,1 mg
Lagerungspuffer RIPA buffer
Reinheit Protein
Testspezifität Homo sapiens GET4, mRNA.
Inhalt und Lagerung -80°C. Avoid freeze-thaw cycles.
Produktlinie Novus
Protein State Native
Zur Verwendung mit (Anwendung) Western Blot

Nur für Forschungszwecke

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.