Learn More
GFAP Antibody (CL2713) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NBP2-76477MFV450
Artikelnummer. 30294481
  • 770.00 € / 0.10 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
GFAP Monoclonal antibody specifically detects GFAP in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
GFAP
Monoclonal
mFluor Violet 450 SE
ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein
This antibody was developed using a recombinant protein derived from P14136, with the exact immunogen sequence remaining proprietary.
0.1 mL
Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers
2670
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
CL2713
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat
Purified