Learn More
Invitrogen™ GGCT Polyclonal Antibody
GREENER_CHOICE
Sheep Polyclonal Antibody
Marke:  Invitrogen™ PA5148958
Artikelnummer. 30533706
  • 429.00 € / 100 Mikrogramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
GGCT (EC 2.3.2.4) catalyzes the formation of 5-oxoproline (pyroglutamic acid) from gamma-glutamyl dipeptides and may play a significant role in glutathione homeostasis.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
GGCT
Polyclonal
Unconjugated
GGCT
A030007L17Rik; AA673177; C7orf24; CRF21; Cytochrome c-releasing factor 21; gamma -glutamyl cyclotransferase; gamma-glutamyl cyclotransferase; gamma-glutamyl cyclotransferase-like 1; gamma-glutamylcyclotransferase; Gctg; GGC; GGCT; Ggctl1; RGD1304876; RGD1562590
Sheep
Antigen affinity chromatography
RUO
79017
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
O75223
GGCT
E. coli-derived recombinant human gamma -Glutamylcyclotransferase/CRF21, Met1-Leu188.