missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ 100mM Glycine
Click to view available options
Menge:
5 L
20 L
50 L
100 L
200 L
Packungsgröße:
100 Liter
20 Liter
200 Liter
5 Liter
50 Liter
Beschreibung
Gibco process buffers are sterile-filtered, packaged in bioprocess containers, and ready to order. Sourcing process buffers can allow you to focus on additional critical steps in your biopharmaceutical processes.
Features
- Available in a range of pack sizes
- Established shelf life; set specifications
- Made to order
- Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
- Readily accessible documentation to help accelerate qualification
Benefits
- Helps ease time requirements for in-house buffer preparation
- Mitigates space constraints for equipment storage
- Streamlines in-house processes related to formulation, hydration, testing, and validation
- Addresses workflow needs for biologics development and manufacturing stages
Additional information
- Multi-compendial testing: compendial-grade or equivalent raw materials
- Primary packaging: Aegis 5-14 single-use bioprocess container (BPC)
- Secondary packaging: rigid shipping container with appropriate dunnage
- Manufacturing facilities: ISO 13485–certified, cGMP-compliant to 21 CFR Part 820 standard
Spezifikation
Spezifikation
| Form | Clear, colorless solution |
| Produkttyp | Glycine |
| Sterilität | Sterile-filtered |
| Sterilization Method | Sterile-filtered |
| Zusammensetzung | Glycine Buffer |
| Fertigungsqualität, Herstellungsqualität | CGMP |
| Menge | 20 L |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur