missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLAST Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA572895
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
GLAST Polyclonal Antibody for Western Blot, IHC (P), IHC (F), ICC/IF, Flow, ELISA
This gene encodes a member of a member of a high affinity glutamate transporter family. Mutations in this gene are associated with episodic ataxia, Type 6. Alternative splicing results in multiple transcript variants.
Spezifikation
| GLAST | |
| Polyclonal | |
| Unconjugated | |
| SLC1A3 | |
| AI504299; B430115D02Rik; EA6; Eaat1; EAAT-1; Excitatory amino acid transporter 1; FLJ25094; GLAST; GLAST1; GLAST-1; Glial glutamate transporter; glial high affinity glutamate transporter; GLU-T; GluT-1; glutamate transporter; glutamate/aspartate transporter; Gmt1; High-affinity neuronal glutamate transporter; MGluT1; Slc1a3; Sodium-dependent glutamate/aspartate transporter 1; solute carrier family 1 (glial high affinity glutamate transporter), member 3; solute carrier family 1 member 3; solute carrier family 1, member 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20512, 29483, 6507 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P24942, P43003, P56564 | |
| SLC1A3 | |
| A synthetic peptide made to a C-terminal portion of the rat SLC1A3 protein (between residues 500-542). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur