Learn More
Glut1 Antibody (GLUT1/2476), Novus Biologicals™
Artikelnummer. 18603368 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
100 μg
20 μg
Packungsgröße:
100 Mikrogramm
20 Mikrogramm
Artikelnummer. Menge unitSize
18603368 100 μg 100 Mikrogramm
18648188 20 μg 20 Mikrogramm
2 options
Artikelnummer. 18603368
Marke: Novus Biologicals NBP275785100ug
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Mouse Monoclonal Antibody
Glut1 Monoclonal antibody specifically detects Glut1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Glut1
Anwendungen Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Klassifikation Monoclonal
Klon GLUT1/2476
Konzentration 0.2 mg/mL
Konjugat Unconjugated
Verdünnung Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, ELISA 1-2 μg/mL, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array
Zusammensetzung 10 mM PBS with 0.05% BSA
Gen-Zugriffsnummer P11166
Gen-Alias Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1
Wirtsspezies Mouse
Immunogen Recombinant fragment of human Glut1 protein (around aa 203-305) (exact sequence is proprietary) (Uniprot: P11166)
Reinigungsverfahren Protein A or G purified
Menge 100 μg
Regulatorischer Status RUO
Forschungsgebiet Cancer, Cardiovascular Biology, Cellular Markers, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroscience, Plasma Membrane Markers, Signal Transduction
Primär oder sekundär Primary
Gen-ID (Entrez) 6513
Zielspezies Human
Inhalt und Lagerung Store at 4°C.
Form Purified
Isotype IgG2b κ