Learn More
Glutathione Peroxidase 1/GPX1 Antibody (3G1), Novus Biologicals™
Rabbit Monoclonal Antibody
Marke:  Novus Biologicals NBP3-26546-100ul
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 30043153
  • 533.00 € / 100 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Glutathione Peroxidase 1/GPX1 Monoclonal antibody specifically detects Glutathione Peroxidase 1/GPX1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Glutathione Peroxidase 1/GPX1
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA
3G1
ELISA
Cellular glutathione peroxidase, EC 1.11.1, EC 1.11.1.9, glutathione peroxidase 1, GPx-1, GSHPx-1, GSHPX1, MGC14399, MGC88245
A synthesized peptide derived from Human Glutathione Peroxidase 1/GPX1 [UniProt P07203]
100 μL
Apoptosis, Cancer, Cell Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
2876
Store at -20 to -70C. Avoid freeze-thaw cycles.