Learn More
PeproTech GM-CSF Polyclonal Antibody, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
Marke: PeproTech 500-P225-100UG
Beschreibung
AA Sequence of recombinant protein: MAPTRSPNPV TRPWKHVDAI KEALSLLNDM RALENEKNED VDIISNEFSI QRPTCVQTRL KLYKQGLRGN LTKLNGALTM IASHYQTNCP PTPETDCEIE VTTFEDFIKN LKGFLFDIPF DCWKPVQK. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat GM-CSF. Anti-Rat GM-CSF-specific antibody was purified by affinity chromatography employing an immobilized Rat GM-CSF matrix. Sandwich ELISA: To detect Rat GM-CSF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Rat GM-CSF (500-P225Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat GM-CSF. Western Blot: To detect Rat GM-CSF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Rat GM-CSF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
The GM-CSF gene encodes for a cytokine that regulates the production, differentiation, and function of macrophages and granulocytes. The active form of the protein exists as a homodimer in the extracellular space. This gene is located in a cluster of related genes at the chromosome region 5q31, which has been associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include interleukins 4, 5, and 13. The gene is involved in promoting tissue inflammation. Elevated levels of cytokines, including the one produced by this gene, have been observed in SARS-CoV-2 infected patients with acute respiratory distress syndrome. Mice that lack this gene or its receptor have been shown to develop pulmonary alveolar proteinosis.
Spezifikation
| GM-CSF | |
| Polyclonal | |
| Unconjugated | |
| CSF2 | |
| colony stimulating factor 2; colony stimulating factor 2 (granulocyte-macrophage); colony-stimulating factor; CSF; CSF2; Csfgm; cytokine; GMCSF; Gm-CSf; granulocyte macrophage colony stimulating factor; granulocyte macrophage-colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGC151255; MGC151257; MGI-IGM; M-GM-CSF; molgramostin; put. GM-CSF; sargramostim | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116630 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P48750 | |
| CSF2 | |
| E.coli-derived Recombinant Rat GM-CSF. | |
| 100 μg | |
| Primary | |
| Rat | |
| Antibody |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.