Learn More
GMPR 1/2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15683-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody can recognize both GMPR1 and GMPR2 due to the high homology.
This gene encodes an enzyme that catalyzes the irreversible and NADPH-dependent reductive deamination of GMP to IMP. The protein also functions in the re-utilization of free intracellular bases and purine nucleosides.Spezifikation
| GMPR 1/2 | |
| Polyclonal | |
| Unconjugated | |
| GMPR | |
| GMP reductase 1, GMPR, GMPR1, GMPR2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 105446, 117533, 192357, 2766, 51292, 66355 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P36959, Q99L27, Q9DCZ1, Q9P2T1, Q9Z244 | |
| GMPR, GMPR2 | |
| GMPR 1/2 Fusion Protein Ag8185 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |