missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Canine IgE Secondary Antibody
Goat Polyclonal Secondary Antibody
Marke: Invitrogen™ SA510257
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody may cross react with IgE from other species.
Anti-Canine secondary antibodies are affinity-purified antibodies with well-characterized specificity for canine (dog) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation
| Canine IgE | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| Canine | |
| Secondary Antibody | |
| IgG |
| ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Canine IgE. | |
| 1 mg | |
| Secondary | |
| 4°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur