Learn More
Goat anti-Chicken IgG (H+L) Secondary Antibody, DyLight 549 (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Marke:  Novus Biologicals NBP1-72814
Artikelnummer. 18572304
  • 258.00 € / 100 Mikrogramm
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Chicken samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Fluorophore-linked immunosorbent assay
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
IgG (H+L)
Polyclonal
DyLight 549
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free
Chicken IgG whole molecule
100 μg
Adaptive Immunity, Immunology
Reconstitute with 100 μg deionized water (or equivalent).
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Western Blot, Immunocytochemistry/Immunofluorescence, Fluorophore-Linked Immunosorbent Assay
LYOPH
Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:5000, Fluorophore-linked immunosorbent assay 1:20000
Goat
Multi-step
RUO
Secondary
Chicken