Learn More
Goat anti-Golden Syrian Hamster IgG (H+L) Secondary Antibody, DyLight 755, Novus Biologicals™
Goat Polyclonal Antibody
Marke:  Novus Biologicals NBP1-72756IR
Artikelnummer. 18484085
  • 312.00 € / 0.50 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Golden Syrian Hamster samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
IgG (H+L)
Polyclonal
50mM Sodium Borate
Golden Syrian Hamster IgG whole molecule
0.5 mL
Adaptive Immunity, Immunology
Golden Syrian Hamster
Purified
Western Blot, ELISA, Immunohistochemistry, Dot Blot
DyLight 755
Goat
Immunogen affinity purified
RUO
Secondary
Store at 4°C in the dark.