missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Hamster IgG (H+L) Secondary Antibody, DyLight 680 (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Marke: Novus Biologicals NBP1-73011
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Hamster samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Spezifikation
| IgG (H+L) | |
| Polyclonal | |
| DyLight 680 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Secondary | |
| Reconstitute with 100 ul deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Western Blot, Immunocytochemistry/Immunofluorescence 1:5000, Fluorophore-linked immunosorbent assay 1:10000 | |
| Armenian and Golden Syrian Hamster IgG, whole molecule | |
| 0.1 mg | |
| Adaptive Immunity, Immunology | |
| This antibody was pre-adsorbed against Mouse and Rat Serum Proteins. No reaction was observed against Mouse or Rat Serum Proteins. This antibody will react with heavy chains of Golden Syrian & Armenian Hamster IgG and with light chains of most Golden Syrian & Armenian Hamster immunoglobulins. | |
| Hamster | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur