missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Human IgG F(ab')2 Secondary Antibody

Artikelnummer. 11835875
Ansicht ändern
Click to view available options
Menge:
2 mg
Packungsgröße:
2 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
11835875 2 mg 2 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11835875 Lieferant Invitrogen™ Lieferanten-Nr. 31122
 Eingeschränkter Artikel
Lager vorrätig
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Goat Polyclonal Secondary Antibody

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31122 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31122 reacts with the light chains of human IgG and with the light chains common to other human immunoglobulins, but not with the Fc portion of human IgG or non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. After opening, dilute only enough antibody for a single day's use. Store remainder at 4°C under sterile conditions. Country of origin: USA.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG F(ab')2
Anwendungen Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Konzentration 2.4 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with no preservative; pH 7.6
Wirtsspezies Goat
Reinigungsverfahren Antigen affinity chromatography
Menge 2 mg
Regulatorischer Status RUO
Primär oder sekundär Secondary
Zielspezies Human
Inhalt und Lagerung 4°C
Produkttyp Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.