missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Human IgG Fab Superclonal™ Secondary Antibody, Alexa Fluor™ Plus 488

Artikelnummer. p-7244472
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Menge:
50 μL
250 μL
missing translation for 'unitSize'
250 Mikroliter
50 Mikroliter
Artikelnummer. Menge unitSize
17854574 250 μL 250 Mikroliter
17854584 50 μL 50 Mikroliter
2 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 17854574

missing translation for 'mfr': Invitrogen™ A56869

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Goat Recombinant Superclonal Secondary Antibody

This Goat-anti-Human IgG Fab, Superclonal Recombinant Secondary Antibody detects human IgG Fab with specificity to the light chains kappa and lambda as well as the heavy chain CH1. This antibody does not detect the Fc and does not cross-react with immunoglobulins from other species. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency. Superclonal Recombinant antibodies are a defined pool of recombinant monoclonal antibodies, manufactured by selecting and pooling individual clones to provide complementary, specific, and optimal epitope coverage to achieve enhanced performance. The sensitivity and specificity of each lot is confirmed using ELISA. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Human IgG Fab
Anwendungen Flow Cytometry, Western Blot, Immunocytochemistry
Klassifikation Recombinant Superclonal
Konzentration 1.0 mg/mL
Konjugat Alexa Fluor Plus 488
Zusammensetzung Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.5
Wirtsspezies Goat
Immunogen Purified Human IgG.
Reinigungsverfahren Affinity chromatography
Menge 250 μL
Regulatorischer Status RUO
Primär oder sekundär Secondary
Zielspezies Human
Inhalt und Lagerung 4°C, store in dark
Produkttyp Secondary Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.