missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Human IgG Fc Secondary Antibody
Goat Polyclonal Secondary Antibody
Marke: Invitrogen™ H10000
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Goat anti-Human IgG Fc Secondary Antibody for Flow
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation
| Human IgG Fc, Goat anti-Human IgG Fc Secondary Antibody | |
| Polyclonal | |
| PBS with 0.1% sodium azide | |
| Purified | |
| ASR | |
| Human | |
| Secondary Antibody | |
| IgG |
| Flow Cytometry | |
| Unconjugated | |
| Goat | |
| 2 mL | |
| Secondary | |
| 4°C, store in dark | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur