missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Human IgG, IgM, IgA (H+L) Secondary Antibody, HRP

Artikelnummer. 11899130
Change view
Click to view available options
Menge:
2 mL
Packungsgröße:
2 Milliliter
Artikelnummer. Menge unitSize
11899130 2 mL 2 Milliliter
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 11899130

Marke: Invitrogen™ 31418

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Goat Polyclonal Secondary Antibody

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31418 has been successfully used in Western blot, and ICC applications. Antibody Specificity: Based on immunoelectrophoresis, the antibody reacts with the heavy chains of human IgA, IgG, IgM and with light chains common to most human immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. However, these antibodies may cross-react with immunoglobulins from other species. Restoration and Storage: Store product at 2-8 °C until opened. Restore with 2.0 mL distilled water (0.8 mg/mL after restoration). Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. To judge clarity, draw product into a pasteur pipette. Product may be stored for several weeks at 2-8 °C as undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Human IgG, IgM, IgA (H+L)
Anwendungen Immunohistochemistry, Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Konzentration 0.8 mg/mL
Konjugat HRP
Zusammensetzung PBS with 15mg/mL BSA and no preservative; pH 7.6
Wirtsspezies Goat
Reinigungsverfahren Antigen affinity chromatography
Menge 2 mL
Regulatorischer Status RUO
Primär oder sekundär Secondary
Zielspezies Human
Inhalt und Lagerung 4°C
Produkttyp Secondary Antibody
Form Lyophilized
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.