Learn More
Invitrogen™ Goat anti-Human Kappa Light Chain Secondary Antibody
Goat Polyclonal Secondary Antibody
Marke: Invitrogen™ SA510285
Beschreibung
This antibody may cross react with kappa light chains from other species.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation
| Human Kappa Light Chain | |
| Polyclonal | |
| Unconjugated | |
| IGHA1 | |
| HCAK1; IgA1; IgG3; IGHA1; IGHA2; IGHG1; IGHG2; IGHG3; IGHG4; IGKC; IGKCD; IGLC; IGLC1; immunoglobulin heavy constant alpha 1; immunoglobulin heavy constant alpha 2 (A2m marker); immunoglobulin heavy constant gamma 1 (G1m marker); immunoglobulin heavy constant gamma 2 (G2m marker); immunoglobulin heavy constant gamma 3 (G3m marker); immunoglobulin heavy constant gamma 4 (G4m marker); immunoglobulin kappa constant; immunoglobulin lambda constant 1; Km | |
| Human Kappa b+f. | |
| 1 mg | |
| Secondary | |
| Human | |
| Secondary Antibody | |
| IgG |
| ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P01834, P01857, P01859, P01860, P01861, P01876, P01877, P0CG04 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 3493, 3494, 3500, 3501, 3502, 3503, 3514, 3537 | |
| 4°C | |
| Liquid |