Learn More
Invitrogen™ Goat anti-Mouse IgG Fc Secondary Antibody
Goat Polyclonal Secondary Antibody
Marke: Invitrogen™ A16086
Beschreibung
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation
| Mouse IgG Fc | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Affinity chromatography | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |
| ELISA | |
| >4.5 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.2 | |
| Gamma Immunoglobin Fc region. | |
| 2 mg | |
| Secondary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.