Learn More
Invitrogen™ Goat anti-Mouse IgG Fc Secondary Antibody, Rhodamine
Goat Polyclonal Secondary Antibody
Marke: Invitrogen™ 31663
Beschreibung
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31663 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31663 reacts with the heavy chains of mouse IgG, but not with the light chains of most mouse immunoglobulins. This antibody does not react against mouse IgM or non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Rhodamine Amax= 550 nm; Emax= 570 nm. Fluorophore/Protein Absorbance Ratio: A550/A280 = ~ 0.4 (lot-dependent). Reconstitute with 1.5 mL of distilled water (1.5 mg/mL after restoration). Country of Origin: USA.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation
| Mouse IgG Fc | |
| Polyclonal | |
| Rhodamine | |
| Goat | |
| 2 mg | |
| Secondary | |
| 4°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
| 1.5 mg/mL | |
| PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 | |
| Affinity chromatography | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.