missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Mouse IgG (H+L) Secondary Antibody, Rhodamine

Artikelnummer. 11849190
Change view
Click to view available options
Menge:
2 mg
Packungsgröße:
2 Milligramm
Artikelnummer. Menge unitSize
11849190 2 mg 2 Milligramm
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 11849190

Marke: Invitrogen™ 31660

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Goat Polyclonal Secondary Antibody

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31660 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31660 reacts with the heavy chains of mouse IgG and with the light chains common to most mouse immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Rhodamine Amax= 550 nm; Emax= 570 nm. Fluorophore/Protein Absorbance Ratio: A550/A280 = ~ 0.4 (lot-dependent). Reconstitute with 1.5 mL of distilled water (1.5 mg/mL after restoration). Country of Origin: USA.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Mouse IgG (H+L)
Anwendungen Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry
Klassifikation Polyclonal
Konzentration 1.5 mg/mL
Konjugat Rhodamine
Zusammensetzung PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6
Wirtsspezies Goat
Reinigungsverfahren Antigen affinity chromatography
Menge 2 mg
Regulatorischer Status RUO
Primär oder sekundär Secondary
Zielspezies Mouse
Inhalt und Lagerung 4°C
Produkttyp Secondary Antibody
Form Lyophilized
Isotype IgG
Mehr anzeigen Weniger anzeigen
compliance-icons
Product Identifier
  • Goat anti-Mouse IgG (H+L) Secondary Antibody, Rhodamine
Signal Word
  • Achtung
Hazard Category
  • Akute Toxizität Kategorie 4
  • LANGFRISTIG GEWÄSSERGEFÄHRDEND Chronisch 3
Hazard Statement
  • H302-Gesundheitsschädlich bei Verschlucken.
  • H412-Schädlich für Wasserorganismen, mit langfristiger Wirkung.
Precautionary Statement
  • P264-Nach Gebrauch gründlich waschen.
  • P270-Bei Gebrauch nicht essen, trinken oder rauchen.
  • P273-Freisetzung in die Umwelt vermeiden.
  • P301+P310-BEI VERSCHLUCKEN: Sofort GIFTINFORMATIONSZENTRUM/Arzt anrufen/
  • P330-Mund ausspülen.
  • P501b-Inhalt/Behälter gemäß den örtlichen/regionalen/nationalen/internationalen Vorschriften zuführen.
Supplemental information
  • MIXTURE LIST-Enthält: SODIUM AZIDE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.