missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Porcine IgG (H+L) Secondary Antibody, HRP, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Spezifikation
Spezifikation
| Antigen | IgG (H+L) |
| Anwendungen | Western Blot, ELISA, Immunohistochemistry |
| Klassifikation | Polyclonal |
| Konzentration | LYOPH |
| Konjugat | HRP |
| Verdünnung | Western Blot 1:1000 - 1:5000, ELISA 1:100000 - 1:200000, Immunohistochemistry 1:500 - 1:2500 |
| Zusammensetzung | Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free |
| Wirtsspezies | Goat |
| Immunogen | Goat anti-Porcine IgG (H+L) Secondary Antibody [HRP] was produced by repeated immunization with Swine IgG whole molecule in goat. |
| Reinigungsverfahren | Multi-step |
| Mehr anzeigen |
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?