Learn More
Invitrogen™ Goat anti-Rabbit IgG (H+L) Secondary Antibody, AP
Artikelnummer. 10406413
Ansicht ändern
Click to view available options
Menge:
1 mL
Packungsgröße:
1 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
10406413 1 mL 1 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Nur f\u00FCr Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.
Artikelnummer. 10406413 Lieferant Invitrogen™ Lieferanten-Nr. 656122
Proteomik vorantreiben
Verwenden Sie den Aktionscode 28003
Alle Werbeaktionen anzeigen
 Eingeschränkter Artikel
Lager vorrätig
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Nur f\u00FCr Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.
Goat Polyclonal Secondary Antibody
Goat anti-Rabbit IgG (H+L) Secondary Antibody for ELISA, IHC (P), IP, Western Blot
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Rabbit IgG (H+L)
Anwendungen ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Klassifikation Polyclonal
Konzentration 1 mg/mL
Konjugat Alkaline Phosphatase (AP)
Zusammensetzung 0.05M tris with 0.15M NaCl, 1mM MgCl2, 1% BSA, 50% glycerol and 0.1% sodium azide; pH 7.5
Wirtsspezies Goat
Immunogen Gamma Immunoglobin.
Reinigungsverfahren Affinity chromatography
Menge 1 mL
Regulatorischer Status RUO
Primär oder sekundär Secondary
Zielspezies Rabbit
Inhalt und Lagerung 4°C
Produkttyp Secondary Antibody
Form Liquid
Isotype IgG