missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Rat IgM (Heavy chain) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 488
Goat Polyclonal Secondary Antibody
Marke: Invitrogen™ A21212
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Goat anti-rat IgM antibodies have been purified by rat IgM-affinity chromatography and react with the mu chain of rat IgM, as determined by ELISA. The rat anti-IgM may also react with IgM from other species. Product will be shipped at Room Temperature.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation
| Rat IgM (Heavy chain) Cross-Adsorbed | |
| Polyclonal | |
| Alexa Fluor 488 | |
| Goat | |
| purified | |
| RUO | |
| Rat | |
| Secondary Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
| 2 mg/mL | |
| PBS with 5mM sodium azide; pH 7.5 | |
| Rat Mu immunonglobin. | |
| 500 μg | |
| Secondary | |
| 4°C, store in dark | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur