Learn More
Gold foil, 0.25mm (0.01in) thick, Premion™, 99.999% (metals basis)
Product Code. 15456585
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456585 50 x 50 mm Each
15446585 25 x 25 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15456585 Supplier Thermo Scientific Alfa Aesar Supplier No. 044163.FI
 Restricted Item
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here
missing translation for 'validMainframeMsg'
CAS: 7440-57-5 | Au | 196.97 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]
Specifications
Quantity 50 x 50 mm
Odor Odorless
Weight ≈3g/25x25mm
Form Foil
Percent Purity 99.999%
Grade Premion
Assay (metals basis)
Chemical Name or Material Gold foil, 0.25mm (0.01 in.) thick