Learn More
Invitrogen™ GP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA542593
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Peptide sequence: SLQAALQPIV SSLNVSVDGN GEFIVRMALF QDQNYTNPYE GDAVELSVES Sequence homology: Cow: 79%; Horse: 92%; Human: 100%; Mouse: 92%; Pig: 79%; Rat: 92%.
GP2 is component of pancreatic secretory (zymogen) granule. The exact function of GP2 remains unknown.
Spezifikation
| GP2 | |
| Polyclonal | |
| Unconjugated | |
| GP2 | |
| 2310037I18Rik; AV060639; glycoprotein 2; glycoprotein 2 (zymogen granule membrane); glycoprotein 80; GP2; gp80; Pancreatic secretory granule membrane major glycoprotein GP2; pancreatic zymogen granule membrane associated protein GP2; pancreatic zymogen granule membrane protein GP-2; secretory (zymogen) granule membrane glycoprotein GP2; ZAP75; zymogen granule membrane glycoprotein 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2813 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P55259 | |
| GP2 | |
| Synthetic peptide directed towards the middle region of human GP2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |