Learn More
Granulin Antibody (CL5695) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NBP2-61425MFV500
Artikelnummer. 30304249
  • 759.00 € / 0.10 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Granulin Monoclonal antibody specifically detects Granulin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Gene Knock-Out
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Granulin
Monoclonal
mFluor Violet 500 SE
acrogranin, GEP, GP88, granulin, granulin-epithelin, granulins, PC cell-derived growth factor, PCDGF, PEPI, PGRN, proepithelin, progranulin
This antibody was developed using a recombinant protein derived from P28799, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cell Cycle and Replication, Lipid and Metabolism
2896
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Gene Knock-Out
CL5695
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen