Learn More
Granzyme B Monoclonal Antibody (16G6), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Marke: Affymetrix eBioscience 14-8822-95
Beschreibung
The 16G6 antibody reacts with mouse Granzyme B (GrB) which is a member of the granzyme serine protease family. GrB is found in the granules of cytotoxic T cells and NK cells. Granzyme B has also been described as CGL1 (cathepsin G-like-1), a serine protease expressed only in cytotoxic T-lymphocytes after cell activation. GrB has been called CTLA-1 (cytotoxic T lymphocyte-associated serine esterase 1) based on identification of mRNA in various cytotoxic T cells, but not observed in non-cytotoxic lymphoid cells. GrB is crucial for the rapid induction of target cell death by apoptosis, induced by interaction with cytotoxic T cells. The receptor involved has been identified as mannose 6-phosphate receptor. This receptor functions as a death receptor for granzyme B during cytotoxic T cell-induced apoptosis. For intracellular staining and flow cytometric analysis with direct conjugates of anti-mouse Granzyme B, it is highly recommended to use the Foxp3 Staining Buffer Set (cat. 00-5523). Other buffers may yield varying results. For more information, please contact technical support at tech@ebioscience.com.
Granzyme B Monoclonal antibody specifically detects Granzyme B in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot
Spezifikation
| Granzyme B | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P04187 | |
| Rat | |
| 10 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b κ |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 16G6 | |
| Unconjugated | |
| GZMB | |
| AI553453; C11; cathepsin G-like 1; CCP1; CCP-1/C11; CCPI; CGL1; CGL-1; CSPB; CSP-B; CTLA1; CTLA-1; CTSGL1; Cytotoxic cell protease 1; cytotoxic serine protease B; cytotoxic T-lymphocyte proteinase 2; cytotoxic T-lymphocyte-associated serine esterase 1; fragmentin; fragmentin 2; fragmentin-2; GLP I; GLP III; GLP-1; granzyme 2; granzyme B; granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1); granzyme B(G,H); Granzyme-2; GranzymeB; granzyme-like protein 1; granzyme-like protein I; GRB; GZB; GZMB; HLP; Human lymphocyte protein; Lymphocyte protease; Natural killer cell protease 1; OTTHUMP00000028189; RNKP-1; SECT; T-cell serine protease 1-3E | |
| Affinity chromatography | |
| RUO | |
| 14939 | |
| 4° C | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.