missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GRCC10 Polyclonal specifically detects GRCC10 in Human samples. It is validated for Immunohistochemistry/Immunohistochemistry (Paraffin).
Specifications
Specifications
| Antigen | GRCC10 |
| Anwendungen | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | APC |
| Verdünnung | Immunohistochemistry-Paraffin |
| Zusammensetzung | PBS |
| Gen-Alias | C10, C10GRCC10, chromosome 12 open reading frame 57, GRCC10, likely ortholog of mouse gene rich cluster, C10, protein C10 |
| Wirtsspezies | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human GRCC10 fragment. |
| Reinigungsverfahren | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?