Learn More
Tocris Bioscience™ GSK 1562590 hydrochloride
Click to view available options
Menge:
10 mg
50 mg
Packungsgröße:
10 Milligramm
50 Milligramm
Beschreibung
GSK 1562590 hydrochloride is a high affinity and selective urotensin II (UT) receptor antagonist (pKi values are 9.14, 9.28, 9.34, 9.64 and 9.66 at monkey, human, mouse, cat and rat recombinant receptors respectively). Exhibits selectivity for UT receptors over a range of GPCRs, ion channels, enzymes and neurotransmitter transporters. Supresses human urotensin-II (hU-II)-induced contraction of isolated rat aorta in vitro and ex vivo. Inhibits the hU-II-induced increase in mean blood pressure in vivo. Orally active.
Spezifikation
Spezifikation
| Menge | 10 mg |
| Produkttyp | GSK 1562590 hydrochloride |
| Reinheit | >98% |
| Molekulargewicht | 617.95 |
| Hemmstoffe | GSK 1562590 hydrochloride |
Nur für Forschungszwecke