Learn More
Tocris Bioscience™ GSK 650394
Beschreibung
GSK 650394 is a serum- and glucocorticoid-regulated kinase 1 (SGK1) inhibitor (IC50 values are 62 and 103 nM for SGK1 and SGK2 respectively). Displays >30-fold selectivity over Akt and other related kinases. Inhibits androgen-stimulated growth of LNCaP cells, a human prostate carcinoma cell line (IC50 ∽ 1 μM).
Spezifikation
Spezifikation
| Menge | 10 mg |
| Produkttyp | GSK 650394 |
| Reinheit | >97% |
| Molekulargewicht | 382.45 |
| Hemmstoffe | GSK 650394 |
Nur für Forschungszwecke