missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Guselkumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA558658
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Guselkumab is a human immunoglobulin G1 lambda (IgG1lambda) monoclonal antibody that selectively blocks interleukin-23. IL-23 is an inflammatory cytokine that activates the CD4+ T-helper (Th17) cell pathway to mediate the inflammatory cascade that induces psoriatic plaque formation 2. In clinical trials, guselkumab demonstrated improved skin clearance and symptomatic improvements in dermatological manifestations of psoriasis. Developed by Janssen, the subcutaneous injection form of guselkumab was approved in July 2017 under the market name Tremfya for the treatment of adult patients with moderate-to-severe plaque psoriasis.
Spezifikation
| Guselkumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CNTO-1959 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur