missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H-2Ld/H-2Db Mouse anti-Mouse, BUV805, Clone: 28-14-8, BD Biosciences™
Mouse Monoclonal Antibody
Marque: BD Biosciences 748416
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
H-2Ld/H-2Db Monoclonal antibody specifically detects H-2Ld/H-2Db in Mouse samples. It is validated for Flow Cytometry
Spécification
| H-2Ld/H-2Db | |
| Monoclonal | |
| Brilliant Ultraviolet 805 | |
| H-2Db/H-2Ld; H2-Db/H2-Ld; Histocompatibility-2 Db/Ld | |
| C3H.SW mouse splenocytes | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| 28-14-8 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV805 under optimal conditions that minimize unconjugated dye and antibody. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu