Learn More
Harmonin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11358-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This gene encodes a scaffold protein that functions in the assembly of Usher protein complexes. The protein contains PDZ domains, a coiled-coil region with a bipartite nuclear localization signal and a PEST degradation sequence. Defects in this gene are the cause of Usher syndrome type 1C and non-syndromic sensorineural deafness autosomal recessive type 18. Multiple transcript variants encoding different isoforms have been found for this gene.Spezifikation
| Harmonin | |
| Polyclonal | |
| Unconjugated | |
| USH1C | |
| AIE 75, AIE75, Antigen NY CO 38/NY CO 37, Harmonin, NY CO 37, NY CO 38, PDZ 45, PDZ 73, PDZ 73/NY CO 38, PDZ73, USH1C, Usher 1C | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10083, 308596, 72088 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9ES64, Q9Y6N9 | |
| Ush1c | |
| Harmonin Fusion Protein Ag1919 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |