Beschreibung
- Simplification of high cell density cell culture fluid clarification unit operations.
- Remove cells, cell debris and DNA
- Optimized for PCV high density CHO cell culture (5-8% PCV)
- Replacement of primary, secondary and guard membrane clarification stages.
- Typical product recovery of 95+% (capsules)
- Synthetic chromatographic harvest media with chemically defined extractables.
- Predictable scaling from discovery to manufacturing.
- Lower total cost of manufacturing compared to centrifugation and depth filtration.
- Lower consumption of buffer and water compared to depth filtration.
- Capsules fit into laboratory to manufacturing scale workflows.
Spezifikation
Spezifikation
| Einwegprodukt | Single-use |
| Filtertyp | Filter Capsule |
| Material (Membran) | PES |
| Sterilität | Non-sterile |
| Menge | 1 Ea. |
| Typ | Chromatography Filter |