missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HE4 Mouse anti-Human, Clone: 5B2D9, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66557-1-IG-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This gene encodes a protein that is a member of the WFDC domain family. The WFDC domain, or WAP Signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor in many family members. This gene is expressed in pulmonary epithelial cells, and was also found to be expressed in some ovarian cancers. The encoded protein is a small secretory protein, which may be involved in sperm maturation.Spezifikation
| HE4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14508 | |
| WFDC2 | |
| HE4 Fusion Protein Ag5936 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| 5B2D9 | |
| Unconjugated | |
| WFDC2 | |
| HE4, WAP5, WFDC2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 10406 | |
| -20°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur