missing translation for 'onlineSavingsMsg'
Learn More

Hepatitis B Virus X antigen Antibody (X36C) - BSA Free, Novus Biologicals™

Product Code. 18743114 Shop All Bio Techne Products
Change view
Click to view available options
Menge:
0.025 mg
Unit Size:
0.03 Milligramm
Product Code. Menge unitSize
18743114 0.025 mg 0.03 Milligramm
1 options
This item is not returnable. View return policy

Product Code. 18743114

Brand: Novus Biologicals NBP2224450.025mg

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Hepatitis B Virus X antigen Monoclonal specifically detects Hepatitis B Virus X antigen in Virus samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis B Virus X antigen
Anwendungen Western Blot, ELISA, Immunoprecipitation
Klassifikation Monoclonal
Klon X36C
Konzentration 1 mg/mL
Konjugat Unconjugated
Verdünnung Western Blot 1 μg/mL, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500
Gen-Alias HBVgp3 X protein HBx, Hepatitis B Virus X Protein, Peptide X, pX, X protein
Gensymbole HBVgp3
Wirtsspezies Mouse
Immunogen Baculovirus expressed recombinant HBx.
Reinigungsverfahren Immunogen affinity purified
Menge 0.025 mg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 944566
Testspezifität This antibody reacts with Virus.
Zielspezies Virus
Inhalt und Lagerung Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.