missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Hepatitis D Virus Large delta Protein Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5117451
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Centrifuge briefly prior to opening.
HDV is the only virus in the genus, Deltaviridae. HDV is not classified into a viral family because it is a unique virus dependent on HBV. HDV is a co-infection of HBV. The envelope of HDV particles contains the Hepatitis B surface antigen (HBsAg). The production and transmission of HDV is entirely dependent on HBV to provide HBsAg. Thus, HDV is considered a satellite virus of HBV. Unlike a classical satellite virus, however, HDV does not share sequence similarity with HBV, and it can replicate independently of HBV. The nucleocapsid is made up of 60 large and small delta antigens. These are the only proteins encoded by HDV.
Spezifikation
| Hepatitis D Virus Large delta Protein | |
| Polyclonal | |
| Unconjugated | |
| HDV | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Hepatitis D virus Large delta protein (HDV genotype I (strain D380)). The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.34 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur