Learn More
Description
Host Species: Mouse
Clone: e2-4001 + 3B5
Isotype: IgG1 + IgG1
Species Reactivity: Human and Rat. Others not known.
Epitope: Cytoplasmic
Immunogen: Cytoplasmic domain of recombinant human HER-2/c-erbB-2 oncoprotein (e2-4001) and a synthetic peptide from the C-terminus of human HER-2 protein (3B5)
Molecular Weight: 185kDa
Positive Control: LS174T cells, or Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (For cytoplasmic)
Specifications
Specifications
| Antigen | HER-2/c-erbB-2/neu Ab-17 |
| Anwendungen | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Klassifikation | Monoclonal |
| Klon | e2-4001 + 3B5 |
| Konzentration | 200μg/mL |
| Konjugat | Unconjugated |
| Wirtsspezies | Mouse |
| Immunogen | Cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein (e2-4001) and a synthetic peptide from the C-terminus of human c-erbB-2 protein (3B5) |
| Menge | 100 μL |
| Regulatorischer Status | ASR, IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.