Learn More
HER2 / ErbB2 / neu / CD340 Mouse, Clone: ERB2/776, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 2064-MSM6-P1
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
| HER2 / ErbB2 / neu / CD340 | |
| Cell Surface | |
| ERB2/776 | |
| Unconjugated | |
| NP_001276865 | |
| Mouse | |
| 185kDa | |
| Protein A/G Purified | |
| RUO | |
| 2064 | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| ERBB2 | |
| Recombinant intracellular domain of human HER2 protein | |
| SKBR-3 Cells or Breast Cancers. | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |